accessibility menu, dialog, popup

UserName

OO(S)O(R)(R)O(S)OO

CAS RN 607-80-7

IUPAC Name: 5-[(1S,3aR,4S,6aR)-4-(2H-1,3-benzodioxol-5-yl)-hexahydrofuro[3,4-c]furan-1-yl]-2H-1,3-benzodioxole
Synonyms: sesamin d-sesamin asarinin D-(+)-sesamin episesamin fagarol sezamin
Molecular Weight (g/mol): 354.36
Molecular Formula: C20H18O6
InChi Key: PEYUIKBAABKQKQ-AFHBHXEDSA-N
SMILES: C1OC2=CC=C(C=C2O1)[C@H]1OC[C@H]2[C@@H]1CO[C@@H]2C1=CC=C2OCOC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)