accessibility menu, dialog, popup

UserName

CAS RN 607-68-1

IUPAC Name: 2,4-dichloroquinazoline
Synonyms: 2,4-dichloroquinazoline
Molecular Weight (g/mol): 199.03
Molecular Formula: C8H4Cl2N2
InChi Key: TUQSVSYUEBNNKQ-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results