accessibility menu, dialog, popup

UserName

CAS RN 60683-30-9

CH3ONCH3H3CNHCH3CH3(E/Z)

CAS RN 60683-30-9

IUPAC Name: N,N'-bis(propan-2-yl)ethoxymethanimidamide
Synonyms: N,N'-diisopropylethoxymethanimidamide
Molecular Weight (g/mol): 172.27
Molecular Formula: C9H20N2O
InChi Key: AUCYHQMYJIEKBO-UHFFFAOYSA-N
SMILES: CCOC(NC(C)C)=NC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results