missing translation for 'onlineSavingsMsg'
Learn More

O-Ethyl-N,N'-diisopropylisourea 98.0+%, TCI America™

Catalog No. E10585ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10585ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10585ML Supplier TCI America Supplier No. E10585ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 60683-30-9
Color Colorless
Boiling Point 65°C
Molecular Formula C9H20N2O
Quantity 5 mL
MDL Number MFCD24554718
Synonym Ethyl N,N′C-Diisopropylcarbamimidate, N,N′C-Diisopropylcarbamimidic Acid Ethyl Ester
InChI Key AUCYHQMYJIEKBO-UHFFFAOYSA-N
SMILES CCOC(NC(C)C)=NC(C)C
IUPAC Name N,N'-bis(propan-2-yl)ethoxymethanimidamide
Molecular Weight (g/mol) 172.27
PubChem CID 20470583
Formula Weight 172.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material O-Ethyl-N,N′-diisopropylisourea
Show More Show Less
TSCA No
Recommended Storage Refrigerator