accessibility menu, dialog, popup

UserName

NHNN

CAS RN 606129-90-2

IUPAC Name: 6-(9H-carbazol-9-yl)-9H-3,9'-bicarbazole
Synonyms: 6-(carbazol-9-yl)-9H-3,9'-bicarbazole
Molecular Weight (g/mol): 497.6
Molecular Formula: C36H23N3
InChi Key: OGDZAJUZGODBKX-UHFFFAOYSA-N
SMILES: N1C2=C(C=C(C=C2)N2C3=C(C=CC=C3)C3=C2C=CC=C3)C2=C1C=CC(=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results