accessibility menu, dialog, popup

UserName

OOOS

CAS RN 6007-85-8

IUPAC Name: 1H,3H-thieno[3,4-c]furan-1,3-dione
Synonyms: thieno[3,4-c]furan-1,3-dione
Molecular Weight (g/mol): 154.14
Molecular Formula: C6H2O3S
InChi Key: XQTUSPVIMZCNPC-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CSC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results