accessibility menu, dialog, popup

UserName

CAS RN 59837-15-9

IUPAC Name: N-[5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-3-yl]acetamide
Synonyms: N-[5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-3-yl]acetamide
Molecular Weight (g/mol): 504.45
Molecular Formula: C20H28N2O13
InChi Key: INMOOBMAIAWVBW-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(OC2=CC=C(C=C2)[N+]([O-])=O)OC(CO)C(O)C1OC1OC(CO)C(O)C(O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results