accessibility menu, dialog, popup

UserName

BHOOH

CAS RN 597554-03-5

IUPAC Name: [10-(naphthalen-2-yl)anthracen-9-yl]boronic acid
Synonyms: 10-(naphthalen-2-yl)anthracen-9-ylboronic acid
Molecular Weight (g/mol): 348.21
Molecular Formula: C24H17BO2
InChi Key: YGVDBZMVEURVOW-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=C(C2=CC=C3C=CC=CC3=C2)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results