accessibility menu, dialog, popup

UserName

CAS RN 59746-40-6

IUPAC Name: 1-(4-methylbenzoyl)pyrrolidine
Synonyms: 1-(4-methylbenzoyl)pyrrolidine
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: QXMOFZGGGOFRPH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results