accessibility menu, dialog, popup

UserName

CAS RN 597-43-3

IUPAC Name: 2,2-dimethylbutanedioic acid
Synonyms: 2,2-dimethylsuccinic acid
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: GOHPTLYPQCTZSE-UHFFFAOYSA-N
SMILES: CC(C)(CC(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)