CAS RN 595-90-4
IUPAC Name:
tetraphenylstannane
Synonyms:
tetraphenyl tin
Molecular Weight (g/mol):
427.13
Molecular Formula:
C24H20Sn
InChi Key:
CRHIAMBJMSSNNM-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Tetraphenyltin 98.0+%, TCI America™
CAS: 595-90-4 Molecular Formula: C24H20Sn Molecular Weight (g/mol): 427.13 MDL Number: MFCD00003006 InChI Key: CRHIAMBJMSSNNM-UHFFFAOYSA-N Synonym: tetraphenyltin,stannane, tetraphenyl,tetraphenyl tin,ccris 6326,tin, tetraphenyl-,,stannane,tetraphenyl,acmc-209mem PubChem CID: 61146 IUPAC Name: tetraphenylstannane SMILES: C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1