CAS RN 594-81-0
IUPAC Name:
2,3-dibromo-2,3-dimethylbutane
Synonyms:
2,3-dibromo-2,3-dimethylbutane
Molecular Weight (g/mol):
243.97
Molecular Formula:
C6H12Br2
InChi Key:
KLFWZEFFWWOMIF-UHFFFAOYSA-N
SMILES:
CC(C)(Br)C(C)(C)Br
1
–
1
of
1
results
1
2,3-Dibromo-2,3-dimethylbutane 98.0+%, TCI America™
CAS: 594-81-0 Molecular Formula: C6H12Br2 Molecular Weight (g/mol): 243.97 MDL Number: MFCD00053168 InChI Key: KLFWZEFFWWOMIF-UHFFFAOYSA-N Synonym: sym-Dibromotetramethylethane, Pinacol Dibromide PubChem CID: 521884 IUPAC Name: 2,3-dibromo-2,3-dimethylbutane SMILES: CC(C)(C(C)(C)Br)Br