accessibility menu, dialog, popup

UserName

CAS RN 592-90-5

IUPAC Name: oxepane
Synonyms: oxepane
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: UHHKSVZZTYJVEG-UHFFFAOYSA-N
SMILES: C1CCCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results