CAS RN 591-55-9
IUPAC Name:
pyrimidin-5-amine
Synonyms:
5-pyrimidinamine
Molecular Weight (g/mol):
95.11
Molecular Formula:
C4H5N3
InChi Key:
FVLAYJRLBLHIPV-UHFFFAOYSA-N
SMILES:
NC1=CN=CN=C1
1
–
1
of
1
results
1
5-Aminopyrimidine 98.0+%, TCI America™
CAS: 591-55-9 Molecular Formula: C4H5N3 Molecular Weight (g/mol): 95.105 MDL Number: MFCD01529870 InChI Key: FVLAYJRLBLHIPV-UHFFFAOYSA-N PubChem CID: 344373 ChEBI: CHEBI:38619 IUPAC Name: pyrimidin-5-amine SMILES: C1=C(C=NC=N1)N