accessibility menu, dialog, popup

UserName

OHH3CNH3CCH3OHO

CAS RN 590-47-6

IUPAC Name: (carboxymethyl)trimethylazanium hydroxide
Synonyms: (carboxymethyl)trimethylammonium hydroxide inner salt
Molecular Weight (g/mol): 135.16
Molecular Formula: C5H13NO3
InChi Key: NJZRLXNBGZBREL-UHFFFAOYSA-N
SMILES: [OH-].C[N+](C)(C)CC(O)=O
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)