accessibility menu, dialog, popup

UserName

CAS RN 58861-53-3

FN

CAS RN 58861-53-3

IUPAC Name: 2-(4-fluorophenyl)pyridine
Synonyms: 2-(4-fluorophenyl)pyridine
Molecular Weight (g/mol): 173.19
Molecular Formula: C11H8FN
InChi Key: MHWIDTQQBWGUCD-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results