accessibility menu, dialog, popup

UserName

H3CNCH3SOOBrNONBr

CAS RN 5874-57-7

IUPAC Name: 4-bromo-N-{1-[3-(4-bromophenyl)-4-oxo-3,4-dihydroquinazolin-2-yl]ethyl}-N-methylbenzene-1-sulfonamide
Synonyms: 4-bromo-N-{1-[3-(4-bromophenyl)-4-oxoquinazolin-2-yl]ethyl}-N-methylbenzenesulfonamide
Molecular Weight (g/mol): 577.29
Molecular Formula: C23H19Br2N3O3S
InChi Key: LYRWWUCOZBTOPL-UHFFFAOYNA-N
SMILES: CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)