accessibility menu, dialog, popup

UserName

CAS RN 58632-95-4

IUPAC Name: tert-butyl (Z)-[cyano(phenyl)methylidene]amino carbonate
Synonyms: tert-butyl (Z)-[cyano(phenyl)methylidene]amino carbonate
Molecular Weight (g/mol): 246.27
Molecular Formula: C13H14N2O3
InChi Key: QQWYQAQQADNEIC-RVDMUPIBSA-N
SMILES: CC(C)(C)OC(=O)O\N=C(/C#N)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)