accessibility menu, dialog, popup

UserName

CH3SO

CAS RN 5834-16-2

IUPAC Name: 3-methylthiophene-2-carbaldehyde
Synonyms: 3-methylthiophene-2-carbaldehyde
Molecular Weight (g/mol): 126.17
Molecular Formula: C6H6OS
InChi Key: BSQKBHXYEKVKMN-UHFFFAOYSA-N
SMILES: CC1=C(SC=C1)C=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)