CAS RN 5820-22-4
IUPAC Name:
[(2-methylprop-2-en-1-yl)oxy]benzene
Synonyms:
[(2-methylprop-2-en-1-yl)oxy]benzene
Molecular Weight (g/mol):
148.21
Molecular Formula:
C10H12O
InChi Key:
LECDNXOCIPRJNJ-UHFFFAOYSA-N
SMILES:
CC(=C)COC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Methallyl phenyl ether, 96%
Methallyl phenyl ether, 96%
CAS: 5820-22-4 Molecular Formula: C10H12O Molecular Weight (g/mol): 148.21 MDL Number: MFCD00008597 InChI Key: LECDNXOCIPRJNJ-UHFFFAOYSA-N Synonym: methallyl phenyl ether,2-methylallyloxy benzene,ether, 2-methylallyl phenyl,benzene, 2-methyl-2-propenyl oxy,2-methylallyl phenyl ether,2-methylprop-2-en-1-yl oxy benzene,2-methyl-2-propen-1-yl oxy benzene,phenyl-methallyl ether,acmc-1b20w PubChem CID: 79893 IUPAC Name: 2-methylprop-2-enoxybenzene SMILES: CC(=C)COC1=CC=CC=C1