CAS RN 580-22-3
IUPAC Name:
quinolin-2-amine
Synonyms:
2-aminoquinoline
Molecular Weight (g/mol):
144.18
Molecular Formula:
C9H8N2
InChi Key:
GCMNJUJAKQGROZ-UHFFFAOYSA-N
SMILES:
NC1=CC=C2C=CC=CC2=N1
1
–
1
of
1
results
1
2-Aminoquinoline 97.0+%, TCI America™
CAS: 580-22-3 Molecular Formula: C9H8N2 Molecular Weight (g/mol): 144.177 MDL Number: MFCD00038079 InChI Key: GCMNJUJAKQGROZ-UHFFFAOYSA-N Synonym: 2-aminoquinoline,2-quinolinamine,quinolinamine,quinoline, 2-amino,quinolin-2-ylamine,2-amino-quinoline,fragment 19,ccris 1677,chembl61236,2-quinolylamine PubChem CID: 11379 IUPAC Name: quinolin-2-amine SMILES: C1=CC=C2C(=C1)C=CC(=N2)N