accessibility menu, dialog, popup

UserName

CAS RN 58-32-2

IUPAC Name: 2-({6-[bis(2-hydroxyethyl)amino]-4,8-bis(piperidin-1-yl)pyrimido[5,4-d][1,3]diazin-2-yl}(2-hydroxyethyl)amino)ethan-1-ol
Synonyms: dipyridamole stenocardil stimolcardio boehringer ingelheim brand of dipyridamole novopharm brand of dipyridamole permiltin antistenocardin curantil miosen curantyl
Molecular Weight (g/mol): 504.64
Molecular Formula: C24H40N8O4
InChi Key: IZEKFCXSFNUWAM-UHFFFAOYSA-N
SMILES: OCCN(CCO)C1=NC(N2CCCCC2)=C2N=C(N=C(N3CCCCC3)C2=N1)N(CCO)CCO
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)