accessibility menu, dialog, popup

UserName

CAS RN 579-39-5

IUPAC Name: bis(4-fluorophenyl)ethane-1,2-dione
Synonyms: bis(4-fluorophenyl)ethane-1,2-dione
Molecular Weight (g/mol): 246.21
Molecular Formula: C14H8F2O2
InChi Key: BRKULQOUSCHDGS-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results