CAS RN 579-18-0
IUPAC Name:
3-benzoylbenzoic acid
Synonyms:
3-benzoylbenzoic acid
Molecular Weight (g/mol):
226.23
Molecular Formula:
C14H10O3
InChi Key:
AXJXRLHTQQONQR-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
3-Benzoylbenzoic acid, 98%
3-Benzoylbenzoic acid, 98%
CAS: 579-18-0 Molecular Formula: C14H10O3 Molecular Weight (g/mol): 226.23 MDL Number: MFCD00002518 InChI Key: AXJXRLHTQQONQR-UHFFFAOYSA-N PubChem CID: 101386 IUPAC Name: 3-benzoylbenzoic acid SMILES: OC(=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1