accessibility menu, dialog, popup

UserName

CAS RN 57744-67-9

IUPAC Name: 6-iodo-2,3-dihydro-1,4-benzodioxine
Synonyms: 6-iodo-2,3-dihydro-1,4-benzodioxine
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: HZTMYTXWFHBHDC-UHFFFAOYSA-N
SMILES: IC1=CC=C2OCCOC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)