accessibility menu, dialog, popup

UserName

CAS RN 5754-18-7

IUPAC Name: 4-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione
Synonyms: 4-methyl-1,2-dihydropyridazine-3,6-dione
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: QAVYOWFNXMHVEL-UHFFFAOYSA-N
SMILES: CC1=CC(=O)NNC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results