accessibility menu, dialog, popup

UserName

CAS RN 575-61-1

IUPAC Name: (3Z)-3-(phenylmethylidene)-1,3-dihydro-2-benzofuran-1-one
Synonyms: benzalphthalide
Molecular Weight (g/mol): 222.24
Molecular Formula: C15H10O2
InChi Key: YRTPZXMEBGTPLM-UVTDQMKNSA-N
SMILES: O=C1O\C(=C/C2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results