accessibility menu, dialog, popup

UserName

CAS RN 57248-14-3

IUPAC Name: 2,5-dichlorothiophene-3-carbonyl chloride
Synonyms: 2,5-dichlorothiophene-3-carbonyl chloride
Molecular Weight (g/mol): 215.47
Molecular Formula: C5HCl3OS
InChi Key: YGDFSMANOSKQRU-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C(Cl)SC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)