accessibility menu, dialog, popup

UserName

CH3NNNCH3

CAS RN 56935-79-6

IUPAC Name: 4-(3,5-dimethyl-1H-pyrazol-1-yl)benzonitrile
Synonyms: 4-(3,5-dimethylpyrazol-1-yl)benzonitrile
Molecular Weight (g/mol): 197.24
Molecular Formula: C12H11N3
InChi Key: BTXLPDNFEZIQBZ-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)