accessibility menu, dialog, popup

UserName

NNO

CAS RN 56671-66-0

IUPAC Name: imidazo[1,5-a]pyridine-3-carbaldehyde
Synonyms: imidazo[1,5-a]pyridine-3-carbaldehyde
Molecular Weight (g/mol): 146.15
Molecular Formula: C8H6N2O
InChi Key: UFDFAMUXKRGTJE-UHFFFAOYSA-N
SMILES: O=CC1=NC=C2C=CC=CN12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results