CAS RN 5662-95-3
IUPAC Name:
8-oxaspiro[4.5]decane-7,9-dione
Synonyms:
8-oxaspiro[4.5]decane-7,9-dione
Molecular Weight (g/mol):
168.19
Molecular Formula:
C9H12O3
InChi Key:
GFWLMILMVMCJDI-UHFFFAOYSA-N
SMILES:
O=C1CC2(CCCC2)CC(=O)O1
1
–
1
of
1
results
1
1,1-Cyclopentanediacetic Anhydride 98.0+%, TCI America™
CAS: 5662-95-3 Molecular Formula: C9H12O3 Molecular Weight (g/mol): 168.19 MDL Number: MFCD00006678 InChI Key: GFWLMILMVMCJDI-UHFFFAOYSA-N Synonym: 8-oxaspiro 4.5 decane-7,9-dione,3,3-tetramethyleneglutaric anhydride,1,1-cyclopentanediacetic anhydride,cyclopentane-1,1-diacetic anhydride,.beta.,.beta.-tetramethyleneglutaric anhydride,acmc-209ltw,1,1-cyclopentanediaceticanhydride,1,1-cyclopentane-diacetic anhydride,8-oxaspiro 4.5 decan-7,9-dione,8-oxaspiro 4.5-7,9-decanedione PubChem CID: 79743 IUPAC Name: 8-oxaspiro[4.5]decane-7,9-dione SMILES: O=C1CC2(CCCC2)CC(=O)O1