accessibility menu, dialog, popup

UserName

CAS RN 565-59-3

CH3CH3CH3H3C

CAS RN 565-59-3

IUPAC Name: 2,3-dimethylpentane
Synonyms: 2,3-dimethylpentane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: WGECXQBGLLYSFP-UHFFFAOYNA-N
SMILES: CCC(C)C(C)C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)