CAS RN 56187-04-3
IUPAC Name:
4-methyl-N-(2-{N-[2-(4-methylbenzenesulfonamido)ethyl]4-methylbenzenesulfonamido}ethyl)benzene-1-sulfonamide
Synonyms:
4-methyl-N-(2-{N-[2-(4-methylbenzenesulfonamido)ethyl]4-methylbenzenesulfonamido}ethyl)benzenesulfonamide
Molecular Weight (g/mol):
565.72
Molecular Formula:
C25H31N3O6S3
InChi Key:
OCDIAWYQMCPHAM-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)NCCN(CCNS(=O)(=O)C1=CC=C(C)C=C1)S(=O)(=O)C1=CC=C(C)C=C1
1
–
1
of
1
results
1
N,N',N″-Tris(p-toluenesulfonyl)diethylenetriamine 98.0+%, TCI America™
CAS: 56187-04-3 Molecular Formula: C25H31N3O6S3 Molecular Weight (g/mol): 565.718 MDL Number: MFCD00015623 InChI Key: OCDIAWYQMCPHAM-UHFFFAOYSA-N Synonym: N,N′C,N′C′C-Tritosyldiethylenetriamine PubChem CID: 316685 IUPAC Name: 4-methyl-N-[2-[(4-methylphenyl)sulfonyl-[2-[(4-methylphenyl)sulfonylamino]ethyl]amino]ethyl]benzenesulfonamide SMILES: CC1=CC=C(C=C1)S(=O)(=O)NCCN(CCNS(=O)(=O)C2=CC=C(C=C2)C)S(=O)(=O)C3=CC=C(C=C3)C