CAS RN 5616-81-9
IUPAC Name:
tert-butyl 2-(methylamino)acetate hydrochloride
Synonyms:
tert-butyl 2-(methylamino)acetate hydrochloride
Molecular Weight (g/mol):
181.66
Molecular Formula:
C7H16ClNO2
InChi Key:
RNLQHMIDSCYLAK-UHFFFAOYSA-N
SMILES:
Cl.CNCC(=O)OC(C)(C)C
1
–
1
of
1
results
1
Promotions Available
Sarcosine tert-butyl ester hydrochloride, 97%
Sarcosine tert-butyl ester hydrochloride, 97%
CAS: 5616-81-9 Molecular Formula: C7H16ClNO2 Molecular Weight (g/mol): 181.66 MDL Number: MFCD00080936 InChI Key: RNLQHMIDSCYLAK-UHFFFAOYSA-N Synonym: h-sar-otbu.hcl,tert-butyl 2-methylamino acetate hydrochloride,sarcosine tert-butyl ester hydrochloride,tert-butyl sarcosinate hydrochloride,tert-butylsarcosinate hydrochloride,h-sar-otbuhcl,sarcosine t-butyl ester hydrochloride,n-methylglycine t-butyl ester hydrochloride,glycine, n-methyl-, 1,1-dimethylethyl ester, hydrochloride,h-sar-otbu hcl PubChem CID: 11252350 IUPAC Name: tert-butyl 2-(methylamino)acetate;hydrochloride SMILES: CC(C)(C)OC(=O)CNC.Cl