accessibility menu, dialog, popup

UserName

BNFFFK

CAS RN 561328-69-6

IUPAC Name: potassium trifluoro(pyridin-3-yl)boranuide
Synonyms: potassium trifluoro(pyridin-3-yl)boranuide
Molecular Weight (g/mol): 185
Molecular Formula: C5H4BF3KN
InChi Key: MNCKCIOWIHHCSC-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CN=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results