accessibility menu, dialog, popup

UserName

CH3CH3CH3N

CAS RN 55897-65-9

IUPAC Name: 2,2-dimethylhexanenitrile
Synonyms: 2,2-dimethylhexanenitrile
Molecular Weight (g/mol): 125.22
Molecular Formula: C8H15N
InChi Key: LXVMQKREAPQFPN-UHFFFAOYSA-N
SMILES: CCCCC(C)(C)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results