accessibility menu, dialog, popup

UserName

CAS RN 557788-37-1

CH3NCH3H3COH3CFFFSOONSOOFFF

CAS RN 557788-37-1

IUPAC Name: ethyl(2-methoxyethyl)dimethylazanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: bistriflylimide anion; ethyl(2-methoxyethyl)dimethylazanium
Molecular Weight (g/mol): 412.36
Molecular Formula: C9H18F6N2O5S2
InChi Key: NGLLWWMHAWYWLY-UHFFFAOYSA-N
SMILES: CC[N+](C)(C)CCOC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results