CAS RN 55715-03-2
IUPAC Name:
4-(bromomethyl)-3-nitrobenzoate
Synonyms:
4-(bromomethyl)-3-nitrobenzoate
Molecular Weight (g/mol):
259.04
Molecular Formula:
C8H5BrNO4
InChi Key:
QMAHVAFURJBOFV-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(CBr)C(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
4-Bromomethyl-3-nitrobenzoic acid, 97%
4-Bromomethyl-3-nitrobenzoic acid, 97%
CAS: 55715-03-2 Molecular Formula: C8H6BrNO4 Molecular Weight (g/mol): 260.043 MDL Number: MFCD00216575 InChI Key: QMAHVAFURJBOFV-UHFFFAOYSA-N Synonym: 4-bromomethyl-3-nitrobenzoic acid,4-bromomethyl-3-nitrobenzoicacid,benzoic acid, 4-bromomethyl-3-nitro,3-nitro-4-bromomethylbenzoic acid,bnba,acmc-1auor,ksc495i9f,3-nitro-4-bromomethyl benzoic acid,4-bromomethyl-3-nitro benzoic acid PubChem CID: 132386 SMILES: C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])CBr