accessibility menu, dialog, popup

UserName

CAS RN 5570-77-4

CH3NCl

CAS RN 5570-77-4

IUPAC Name: 4-chloro-1-methylpiperidine
Synonyms: 4-chloro-1-methylpiperidine
Molecular Weight (g/mol): 133.62
Molecular Formula: C6H12ClN
InChi Key: MYGXGCCFTPKWIH-UHFFFAOYSA-N
SMILES: CN1CCC(Cl)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results