accessibility menu, dialog, popup

UserName

CAS RN 557-93-7

IUPAC Name: 2-bromoprop-1-ene
Synonyms: 1-propene, 2-bromo-
Molecular Weight (g/mol): 120.98
Molecular Formula: C3H5Br
InChi Key: PHMRPWPDDRGGGF-UHFFFAOYSA-N
SMILES: CC(Br)=C
Boiling Point 
  • (3)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)