CAS RN 5565-32-2
IUPAC Name:
3-{[(5E)-4-oxo-3-phenyl-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}phenyl furan-2-carboxylate
Synonyms:
3-{[(5E)-4-oxo-3-phenyl-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}phenyl furan-2-carboxylate
Molecular Weight (g/mol):
407.46
Molecular Formula:
C21H13NO4S2
InChi Key:
CVCSUQKXXAAHNH-QGOAFFKASA-N
SMILES:
O=C(OC1=CC=CC(\C=C2\SC(=S)N(C2=O)C2=CC=CC=C2)=C1)C1=CC=CO1
1
–
1
of
1
results
1
Chlorotris(trimethylsilyl)silane 95.0+%, TCI America™
CAS: 5565-32-2 Molecular Formula: C21H13NO4S2 Molecular Weight (g/mol): 407.46 MDL Number: MFCD01321222 InChI Key: CVCSUQKXXAAHNH-QGOAFFKASA-N Synonym: Tris(trimethylsilyl)silyl Chloride PubChem CID: 4641097 IUPAC Name: 3-{[(5E)-4-oxo-3-phenyl-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}phenyl furan-2-carboxylate SMILES: O=C(OC1=CC=CC(\C=C2\SC(=S)N(C2=O)C2=CC=CC=C2)=C1)C1=CC=CO1