accessibility menu, dialog, popup

UserName

BrNOOO

CAS RN 5551-12-2

IUPAC Name: 4-bromo-2-nitrobenzaldehyde
Synonyms: 4-bromo-2-nitrobenzaldehyde
Molecular Weight (g/mol): 230.02
Molecular Formula: C7H4BrNO3
InChi Key: GSXUXSXBEUJRAJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=O)C=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results