accessibility menu, dialog, popup

UserName

CAS RN 5525-40-6

ON

CAS RN 5525-40-6

IUPAC Name: 2,4-diphenyl-5,6,7,8-tetrahydroquinolin-5-one
Synonyms: 2,4-diphenyl-7,8-dihydro-6H-quinolin-5-one
Molecular Weight (g/mol): 299.37
Molecular Formula: C21H17NO
InChi Key: PHBAXBAJPLNXRV-UHFFFAOYSA-N
SMILES: O=C1CCCC2=NC(=CC(C3=CC=CC=C3)=C12)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results