accessibility menu, dialog, popup

UserName

H3CN

CAS RN 552-82-9

IUPAC Name: N-methyl-N-phenylaniline
Synonyms: diphenyl methylamine
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: DYFFAVRFJWYYQO-UHFFFAOYSA-N
SMILES: CN(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)