accessibility menu, dialog, popup

UserName

CAS RN 546122-71-8

IUPAC Name: bromo[(2,4-difluorophenyl)methyl]magnesium
Synonyms: bromo[(2,4-difluorophenyl)methyl]magnesium
Molecular Weight (g/mol): 231.32
Molecular Formula: C7H5BrF2Mg
InChi Key: BPKTYCGKUHSSCG-UHFFFAOYSA-M
SMILES: FC1=CC(F)=C(C[Mg]Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results