CAS RN 5459-58-5
CAS RN 5459-58-5
IUPAC Name:
butyl 2-cyanoacetate
Synonyms:
butyl 2-cyanoacetate
Molecular Weight (g/mol):
141.17
Molecular Formula:
C7H11NO2
InChi Key:
DJACTCNGCHPGOI-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)CC#N
1
–
1
of
1
results
1
Butyl Cyanoacetate 98.0+%, TCI America™
CAS: 5459-58-5 Molecular Formula: C7H11NO2 Molecular Weight (g/mol): 141.17 MDL Number: MFCD00001941 InChI Key: DJACTCNGCHPGOI-UHFFFAOYSA-N Synonym: butyl cyanoacetate,n-butyl cyanoacetate,acetic acid, cyano-, butyl ester,unii-f9h6l20voh,cyanoacetic acid butyl ester,cyanoacetic acid n-butyl ester,acetic acid, 2-cyano-, butyl ester,f9h6l20voh,butyl 2-cyanoethanoate,acmc-209lho PubChem CID: 79571 IUPAC Name: butyl 2-cyanoacetate SMILES: CCCCOC(=O)CC#N