CAS RN 5445-30-7
IUPAC Name:
1-butylcyclohexan-1-ol
Synonyms:
1-butylcyclohexan-1-ol
Molecular Weight (g/mol):
156.27
Molecular Formula:
C10H20O
InChi Key:
RCHLXMOXBJRGNX-UHFFFAOYSA-N
SMILES:
CCCCC1(O)CCCCC1
1
–
1
of
1
results
1
Promotions Available
1-n-Butylcyclohexanol, 98%
1-n-Butylcyclohexanol, 98%
CAS: 5445-30-7 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.269 MDL Number: MFCD00021404 InChI Key: RCHLXMOXBJRGNX-UHFFFAOYSA-N Synonym: 1-butylcyclohexanol,cyclohexanol, 1-butyl,1-n-butylcyclohexanol,butylcyclohexanol,1-butyl-cyclohexanol,1-butyl-cyclohexan-1-ol PubChem CID: 138505 IUPAC Name: 1-butylcyclohexan-1-ol SMILES: CCCCC1(CCCCC1)O