accessibility menu, dialog, popup

UserName

CH3OONHOOOOOH

CAS RN 54375-47-2

IUPAC Name: 2-[(carboxylatomethyl)[(7-hydroxy-4-methyl-2-oxo-2H-chromen-8-yl)methyl]azaniumyl]acetate
Synonyms: 2-[(carboxylatomethyl)[(7-hydroxy-4-methyl-2-oxochromen-8-yl)methyl]ammonio]acetate
Molecular Weight (g/mol): 320.28
Molecular Formula: C15H14NO7
InChi Key: RJPSHDMGSVVHFA-UHFFFAOYSA-M
SMILES: CC1=CC(=O)OC2=C(C[NH+](CC([O-])=O)CC([O-])=O)C(O)=CC=C12
Color 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results