accessibility menu, dialog, popup

UserName

CAS RN 5434-30-0

IUPAC Name: N-(5-amino-2-methylphenyl)acetamide
Synonyms: N-(5-amino-2-methylphenyl)acetamide
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: UAZGSMMESOKKQZ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(N)=CC=C1C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)