accessibility menu, dialog, popup

UserName

H3CCH3Br

CAS RN 5433-01-2

IUPAC Name: 1-bromo-3-(propan-2-yl)benzene
Synonyms: 1-bromo-3-isopropylbenzene
Molecular Weight (g/mol): 199.09
Molecular Formula: C9H11Br
InChi Key: GBSGGFCCQZUXNB-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results